Learn More
Selleck Chemical LLC Saikosaponin D S5454-1mg
SDP
Catalog No. 503108597
Encompass_Preferred
Saikosaponin D a calcium mobilizing agent (SERCA inhibitor) is a type of Saponin derivative which is a component extracted from Bupleurum falactum Saikosaponin D has anti-cancer activities
Catalog No. 50-310-8597 Supplier Selleck Chemical LLC Supplier No. S54541MG
Saikosaponin D a calcium mobilizing agent (SERCA inhibitor) is a type of Saponin derivative which is a component extracted from Bupleurum falactum Saikosaponin D has anti-cancer activities