Learn More
Cayman Chemical SaIkosaponIn Dhydrate 10mg
SDP
Catalog No. 502946352
Encompass_Preferred
A triterpene saponin with diverse biological activities; inhibits the proliferation of, and induces apoptosis in, SW1736, ARO, and 8305C anaplastic thyroid cancer cells from 5-20 µM; reduces tumor weight and volume in an ARO mouse xenograft model at 5, 10, and 20 mg/kg; reverses reductions in gut microbial diversity, reduces colonic inflammatory cell infiltration, mucosal thickening, and goblet cell depletion, and increases colon length in a mouse model of DSS-induced ulcerative colitis; has antidepressant-like activity and inhibits LPS-induced hippocampal microglial activation in mice at 1 mg/kg
Catalog No. 50-294-6352 Supplier Cayman Chemical Supplier No. 3315610 MG
A triterpene saponin with diverse biological activities; inhibits the proliferation of, and induces apoptosis in, SW1736, ARO, and 8305C anaplastic thyroid cancer cells from 5-20 µM; reduces tumor weight and volume in an ARO mouse xenograft model at 5, 10, and 20 mg/kg; reverses reductions in gut microbial diversity, reduces colonic inflammatory cell infiltration, mucosal thickening, and goblet cell depletion, and increases colon length in a mouse model of DSS-induced ulcerative colitis; has antidepressant-like activity and inhibits LPS-induced hippocampal microglial activation in mice at 1 mg/kg