Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin E | 64340-44-9 | MFCD32197349 | 99.8% | 764.98 g·mol⁻¹ | C42H68O12 | 10 MG
SDP

Catalog No. 5000289925
Encompass_Preferred

Saikosaponin E is a triterpene saponin isolated from Bupleurum species and provided as a research-grade chemical reagent for biochemical and pharmacological studies. It is supplied as a white to off-white solid, with documented solubility and storage conditions and supporting analytical documents.

  • High purity: 99.8% (HPLC), suitable for research applications.
  • Well characterized: COA, HNMR, MS, and RP-HPLC data available.
  • Solubility: DMSO 100 mg/mL (ultrasonic recommended); in vivo formulations ≥2.5 mg/mL.
  • Storage: solid at 4°C protected from light; in solution -80°C (6 months) or -20°C (1 month).
  • Application: suitable for in vitro and in vivo pharmacology and biochemical studies.

Catalog No. 50-002-89925 Supplier Medchemexpress LLC Supplier No. HYN421110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin E is a triterpene saponin isolated from Bupleurum species and provided as a research-grade chemical reagent for biochemical and pharmacological studies. It is supplied as a white to off-white solid, with documented solubility and storage conditions and supporting analytical documents.

  • High purity: 99.8% (HPLC), suitable for research applications.
  • Well characterized: COA, HNMR, MS, and RP-HPLC data available.
  • Solubility: DMSO 100 mg/mL (ultrasonic recommended); in vivo formulations ≥2.5 mg/mL.
  • Storage: solid at 4°C protected from light; in solution -80°C (6 months) or -20°C (1 month).
  • Application: suitable for in vitro and in vivo pharmacology and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.