Learn More
Medchemexpress LLC Saikosaponin F | 62687-63-2 | 5 MG
SDP
Catalog No. 5000420884
Encompass_Preferred
Saikosaponin F is a component found in Bupleurum (B.) falcatum L. It has a molecular weight of 929.14 and a chemical formula of C48H80O17. This high-purity compound (99.90%) appears as a white to off-white solid and is classified under terpenoids, specifically triterpenes. It is derived from plants such as Compositae and Umbelliferae.
  • Research-grade compound
  • High purity (99.90%)
  • Naturally derived from plants
  • Classified as a triterpene
  • Available in solid form
Catalog No. 50-004-20884 Supplier Medchemexpress LLC Supplier No. HYN21785MG
Saikosaponin F is a component found in Bupleurum (B.) falcatum L. It has a molecular weight of 929.14 and a chemical formula of C48H80O17. This high-purity compound (99.90%) appears as a white to off-white solid and is classified under terpenoids, specifically triterpenes. It is derived from plants such as Compositae and Umbelliferae.
  • Research-grade compound
  • High purity (99.90%)
  • Naturally derived from plants
  • Classified as a triterpene
  • Available in solid form