Learn More
Medchemexpress LLC Saikosaponin F | 62687-63-2 | MFCD32005270 | 99.9% | 929.14 | C48H80O17 | 10 MG
SDP
Catalog No. 5000295878
Encompass_Preferred
Saikosaponin F is a naturally occurring triterpene saponin isolated from Bupleurum falcatum L., supplied for research use. It is a white to off-white solid with high purity and is used in biochemical and pharmacological studies of natural products.
  • High purity (~99.9%), suitable for research applications.
  • Molecular formula C48H80O17; molecular weight 929.14.
  • White to off-white solid, easy to handle and dissolve in suitable solvents.
  • Store sealed, away from moisture and light; recommended storage at 4°C.
  • Documentation available: datasheet, COA, and SDS for quality and safety information.
Catalog No. 50-002-95878 Supplier Medchemexpress LLC Supplier No. HYN217810MG
Saikosaponin F is a naturally occurring triterpene saponin isolated from Bupleurum falcatum L., supplied for research use. It is a white to off-white solid with high purity and is used in biochemical and pharmacological studies of natural products.
  • High purity (~99.9%), suitable for research applications.
  • Molecular formula C48H80O17; molecular weight 929.14.
  • White to off-white solid, easy to handle and dissolve in suitable solvents.
  • Store sealed, away from moisture and light; recommended storage at 4°C.
  • Documentation available: datasheet, COA, and SDS for quality and safety information.