Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saikosaponin G | 99365-19-2 | 1 MG
SDP

Catalog No. 5000420576
Encompass_Preferred

Saikosaponin G is a triterpene glycoside isolated from Bupleurum chinensis. It has a purity of 98.67% and is for research use only.

  • Molecular weight: 780.98
  • Formula: C₄₂H₆₈O₁₃
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO: 50 mg/mL (64.02 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-004-20576 Supplier Medchemexpress LLC Supplier No. HYN42161MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saikosaponin G is a triterpene glycoside isolated from Bupleurum chinensis. It has a purity of 98.67% and is for research use only.

  • Molecular weight: 780.98
  • Formula: C₄₂H₆₈O₁₃
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO: 50 mg/mL (64.02 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.