Learn More
Pacer Global LLC Saint Gobain 072 C-Flex TPE Tubing, Opaque, 0.063'' ID, 0.188'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA
SDP
Catalog No. 5000332298
Encompass_Preferred
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.063" ID, 0.188" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32298 Supplier Pacer Global LLC Supplier No. MLS0720632
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.063" ID, 0.188" OD, 0.0625" Wall, 50 Foot Coil, 1/EA