Learn More
Pacer Global LLC Saint Gobain 072 C-Flex TPE Tubing, Opaque, 0.5'' ID, 0.688'' OD, 0.094'' Wall, 50 Foot Coil, 1/EA
SDP
Catalog No. 5000332233
Encompass_Preferred
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.5" ID, 0.688" OD, 0.094" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32233 Supplier Pacer Global LLC Supplier No. MLS0725003
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.5" ID, 0.688" OD, 0.094" Wall, 50 Foot Coil, 1/EA