Learn More
Pacer Global LLC Saint Gobain 072 C-Flex TPE Tubing, Opaque, 0.625'' ID, 0.875'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA
SDP
Catalog No. 5000332452
Encompass_Preferred
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32452 Supplier Pacer Global LLC Supplier No. MLS0726254
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA