Learn More
Pacer Global LLC Saint Gobain 072 C-Flex TPE Tubing, Opaque, 0.75'' ID, 1.125'' OD, 0.1875'' Wall, 15 Foot Coil, 1/EA
SDP
Catalog No. 5000332299
Encompass_Preferred
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.75" ID, 1.125" OD, 0.1875" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32299 Supplier Pacer Global LLC Supplier No. MLS0727506
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 0.75" ID, 1.125" OD, 0.1875" Wall, 15 Foot Coil, 1/EA