Learn More
Pacer Global LLC Saint Gobain 072 C-Flex TPE Tubing, Opaque, 1'' ID, 1.375'' OD, 0.1875'' Wall, 15 Foot Coil, 1/EA
SDP
Catalog No. 5000332396
Encompass_Preferred
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 1" ID, 1.375" OD, 0.1875" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32396 Supplier Pacer Global LLC Supplier No. MLS07210006
Saint-Gobain - 072 C-Flex TPE Tubing, Opaque, 1" ID, 1.375" OD, 0.1875" Wall, 15 Foot Coil, 1/EA