Learn More
Pacer Global LLC Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.625'' ID, 1'' OD, 0.1875'' Wall, 15 Foot Coil, 1/EA - ACC40048
SDP
Catalog No. 5000332345
Encompass_Preferred
Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.625" ID, 1" OD, 0.1875" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32345 Supplier Pacer Global LLC Supplier No. MLSACC40048
Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.625" ID, 1" OD, 0.1875" Wall, 15 Foot Coil, 1/EA