Learn More
Pacer Global LLC Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.75'' ID, 1.125'' OD, 0.1875'' Wall, 15 Foot Coil, 1/EA - ACC40055
SDP
Catalog No. 5000332458
Encompass_Preferred
Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.75" ID, 1.125" OD, 0.1875" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32458 Supplier Pacer Global LLC Supplier No. MLSACC40055
Saint-Gobain - 082 BrC-Flex TPE Tubing, 0.75" ID, 1.125" OD, 0.1875" Wall, 15 Foot Coil, 1/EA