Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.125'' ID, 0.25'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - 082-125-2
SDP
Catalog No. 5000332453
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32453 Supplier Pacer Global LLC Supplier No. MLS0821252
Saint-Gobain - 082 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 50 Foot Coil, 1/EA