Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.188'' ID, 0.313'' OD, 0.0625'' Wall, 350 Foot Coil, 1/EA - 082-188-2M
SDP
Catalog No. 5000332261
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.188" ID, 0.313" OD, 0.0625" Wall, 350 Foot Coil, 1/EA
Catalog No. 50-003-32261 Supplier Pacer Global LLC Supplier No. MLS0821882M
Saint-Gobain - 082 C-Flex TPE Tubing, 0.188" ID, 0.313" OD, 0.0625" Wall, 350 Foot Coil, 1/EA