Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.25'' ID, 0.375'' OD, 0.0625'' Wall, 1000 Foot Coil, 1/EA - 082-250-2S
SDP
Catalog No. 5000332399
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 1000 Foot Coil, 1/EA
Catalog No. 50-003-32399 Supplier Pacer Global LLC Supplier No. MLS0822502S
Saint-Gobain - 082 C-Flex TPE Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 1000 Foot Coil, 1/EA