Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.375'' ID, 0.5'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - 082-375-2
SDP
Catalog No. 5000332307
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.5" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32307 Supplier Pacer Global LLC Supplier No. MLS0823752
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.5" OD, 0.0625" Wall, 50 Foot Coil, 1/EA