Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.375'' ID, 0.564'' OD, 0.0945'' Wall, 200 Foot Coil, 1/EA - 082-375-3M
SDP
Catalog No. 5000332308
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.564" OD, 0.0945" Wall, 200 Foot Coil, 1/EA
Catalog No. 50-003-32308 Supplier Pacer Global LLC Supplier No. MLS0823753M
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.564" OD, 0.0945" Wall, 200 Foot Coil, 1/EA