Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.375'' ID, 0.625'' OD, 0.125'' Wall, 350 Foot Coil, 1/EA - 082-375-4S
SDP
Catalog No. 5000332309
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.625" OD, 0.125" Wall, 350 Foot Coil, 1/EA
Catalog No. 50-003-32309 Supplier Pacer Global LLC Supplier No. MLS0823754S
Saint-Gobain - 082 C-Flex TPE Tubing, 0.375" ID, 0.625" OD, 0.125" Wall, 350 Foot Coil, 1/EA