Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.625'' ID, 0.875'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA - 082-625-4
SDP
Catalog No. 5000332356
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32356 Supplier Pacer Global LLC Supplier No. MLS0826254
Saint-Gobain - 082 C-Flex TPE Tubing, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA