Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 1'' ID, 1.375'' OD, 0.1875'' Wall, 15 Foot Coil, 1/EA - 082-1000-6
SDP
Catalog No. 5000332466
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 1" ID, 1.375" OD, 0.1875" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32466 Supplier Pacer Global LLC Supplier No. MLS08210006
Saint-Gobain - 082 C-Flex TPE Tubing, 1" ID, 1.375" OD, 0.1875" Wall, 15 Foot Coil, 1/EA