Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.063'' ID, 0.125'' OD, 0.031'' Wall, 50 Foot Coil, 1/EA - 374-063-1
SDP
Catalog No. 5000332305
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.063" ID, 0.125" OD, 0.031" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32305 Supplier Pacer Global LLC Supplier No. MLS3740631
Saint-Gobain - 374 C-Flex TPE Tubing, 0.063" ID, 0.125" OD, 0.031" Wall, 50 Foot Coil, 1/EA