Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.125'' ID, 0.25'' OD, 0.0625'' Wall, 500 Foot Coil, 1/EA - 374-125-2M
SDP
Catalog No. 5000332219
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 500 Foot Coil, 1/EA
Catalog No. 50-003-32219 Supplier Pacer Global LLC Supplier No. MLS3741252M
Saint-Gobain - 374 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 500 Foot Coil, 1/EA