Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.188'' ID, 0.313'' OD, 0.0625'' Wall, 350 Foot Coil, 1/EA - 374-188-2M
SDP
Catalog No. 5000332239
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.188" ID, 0.313" OD, 0.0625" Wall, 350 Foot Coil, 1/EA
Catalog No. 50-003-32239 Supplier Pacer Global LLC Supplier No. MLS3741882M
Saint-Gobain - 374 C-Flex TPE Tubing, 0.188" ID, 0.313" OD, 0.0625" Wall, 350 Foot Coil, 1/EA