Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.188'' ID, 0.375'' OD, 0.0935'' Wall, 350 Foot Coil, 1/EA - 374-188-3M
SDP
Catalog No. 5000332469
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.188" ID, 0.375" OD, 0.0935" Wall, 350 Foot Coil, 1/EA
Catalog No. 50-003-32469 Supplier Pacer Global LLC Supplier No. MLS3741883M
Saint-Gobain - 374 C-Flex TPE Tubing, 0.188" ID, 0.375" OD, 0.0935" Wall, 350 Foot Coil, 1/EA