Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.25'' ID, 0.375'' OD, 0.0625'' Wall, 350 Foot Coil, 1/EA - 374-250-2M
SDP
Catalog No. 5000332455
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 350 Foot Coil, 1/EA
Catalog No. 50-003-32455 Supplier Pacer Global LLC Supplier No. MLS3742502M
Saint-Gobain - 374 C-Flex TPE Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 350 Foot Coil, 1/EA