Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.25'' ID, 0.438'' OD, 0.094'' Wall, 700 Foot Coil, 1/EA - 374-250-3S
SDP
Catalog No. 5000332405
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.25" ID, 0.438" OD, 0.094" Wall, 700 Foot Coil, 1/EA
Catalog No. 50-003-32405 Supplier Pacer Global LLC Supplier No. MLS3742503S
Saint-Gobain - 374 C-Flex TPE Tubing, 0.25" ID, 0.438" OD, 0.094" Wall, 700 Foot Coil, 1/EA