Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 1'' ID, 1.5'' OD, 0.25'' Wall, 15 Foot Coil, 1/EA - 374-1000-8
SDP
Catalog No. 5000332342
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 1" ID, 1.5" OD, 0.25" Wall, 15 Foot Coil, 1/EA
Catalog No. 50-003-32342 Supplier Pacer Global LLC Supplier No. MLS37410008
Saint-Gobain - 374 C-Flex TPE Tubing, 1" ID, 1.5" OD, 0.25" Wall, 15 Foot Coil, 1/EA