Learn More
Accuflow Systems Saint Gobain BarbLock, 0.375 x 0.625 in., Polypropylene, 1 ea
Catalog No. NC3936873
Encompass_Preferred
The BarbLock® retainer is designed for retaining flexible tubing and hose onto barbed fittings. It provides full 360° radial crimp and compression over the barb's ridge, addressing leak path problems. Made from high-performance polypropylene (PP) and PVDF resins, these retainers meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements and do not require adhesives or solvents.
  • Compatible with all flexible tubing materials.
  • Provides full 360° radial crimp behind the barb and 360° compression directly on the barb's ridge.
  • Eliminates the need for solvent bonding, ultraviolet bonding, and toxic adhesives.
  • Manufactured from resins that meet USP <88> Class VI, USP <87>, and/or ISO 10993-5.
  • Offers custom color and identification options.
  • Quick and easy to assemble.
  • PP and PVDF BarbLocks are gamma stable.
Catalog No. NC3936873 Supplier Accuflow Systems Supplier No. BL135377ALRR
The BarbLock® retainer is designed for retaining flexible tubing and hose onto barbed fittings. It provides full 360° radial crimp and compression over the barb's ridge, addressing leak path problems. Made from high-performance polypropylene (PP) and PVDF resins, these retainers meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements and do not require adhesives or solvents.
  • Compatible with all flexible tubing materials.
  • Provides full 360° radial crimp behind the barb and 360° compression directly on the barb's ridge.
  • Eliminates the need for solvent bonding, ultraviolet bonding, and toxic adhesives.
  • Manufactured from resins that meet USP <88> Class VI, USP <87>, and/or ISO 10993-5.
  • Offers custom color and identification options.
  • Quick and easy to assemble.
  • PP and PVDF BarbLocks are gamma stable.