Learn More
Accuflow Systems Saint-Gobain BarbLock Assembly Hand Tool, 187 Series, Steel, 1 ea
Catalog No. NC3938124
Encompass_Preferred
This hand tool is engineered for low volume assembly applications involving 187 series components. Constructed from PVDF and stainless steel, it provides a secure and durable solution for tubing retention tasks.
  • Designed for low volume assembly jobs
  • Constructed from durable PVDF and steel materials
  • Compatible with 187 series tubing with a 0.187 in. inside diameter
  • Engineered for a tubing outside diameter of 0.312 in.
Catalog No. NC3938124 Supplier Accuflow Systems Supplier No. BLT135187
This hand tool is engineered for low volume assembly applications involving 187 series components. Constructed from PVDF and stainless steel, it provides a secure and durable solution for tubing retention tasks.
  • Designed for low volume assembly jobs
  • Constructed from durable PVDF and steel materials
  • Compatible with 187 series tubing with a 0.187 in. inside diameter
  • Engineered for a tubing outside diameter of 0.312 in.