Learn More
Accuflow Systems BarbLock Hand Tool, 500 Series, Stainless Steel, 1 ea
Catalog No. NC3938132
Encompass_Preferred
This 500 Series hand tool is designed for the secure assembly of tubing retainers onto barbed fittings. It provides an advanced alternative to traditional cable ties or clamps, ensuring a consistent and reliable connection for flexible tubing in fluid transfer applications. The tool facilitates a 360° radial crimp, effectively eliminating common leak paths without the need for adhesives or solvents.
  • Provides complete 360° compression over the barb ridge for leak-free connections.
  • Compatible with all flexible tubing materials.
  • Well-suited for systems handling sensitive liquids and gases.
  • Quick and easy to assemble.
Catalog No. NC3938132 Supplier Accuflow Systems Supplier No. BLT135500
This 500 Series hand tool is designed for the secure assembly of tubing retainers onto barbed fittings. It provides an advanced alternative to traditional cable ties or clamps, ensuring a consistent and reliable connection for flexible tubing in fluid transfer applications. The tool facilitates a 360° radial crimp, effectively eliminating common leak paths without the need for adhesives or solvents.
  • Provides complete 360° compression over the barb ridge for leak-free connections.
  • Compatible with all flexible tubing materials.
  • Well-suited for systems handling sensitive liquids and gases.
  • Quick and easy to assemble.