Learn More
Accuflow Systems Saint Gobain BarbLock, PP, 0.500 by 0.750 in., 1 ea
Catalog No. NC3935998
Encompass_Preferred
The BarbLock® retainer securely retains flexible tubing and hose onto barbed fittings. It features a full 360° radial crimp behind the barb and provides 360° of compression directly over the barb's ridge, addressing leak path problems in demanding fluid transfer applications. The one-piece AL series retainer simplifies assembly steps and reduces potential errors.
  • Compatible with all flexible tubing materials.
  • Full 360° radial crimp behind the barb.
  • Provides 360° compression directly on the barb's ridge.
  • Eliminates solvent bonding, UV bonding, and toxic adhesives.
  • Manufactured from resins meeting USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements.
  • Quick and easy to assemble.
  • Gamma stable.
Catalog No. NC3935998 Supplier Accuflow Systems Supplier No. BL135500AL
The BarbLock® retainer securely retains flexible tubing and hose onto barbed fittings. It features a full 360° radial crimp behind the barb and provides 360° of compression directly over the barb's ridge, addressing leak path problems in demanding fluid transfer applications. The one-piece AL series retainer simplifies assembly steps and reduces potential errors.
  • Compatible with all flexible tubing materials.
  • Full 360° radial crimp behind the barb.
  • Provides 360° compression directly on the barb's ridge.
  • Eliminates solvent bonding, UV bonding, and toxic adhesives.
  • Manufactured from resins meeting USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements.
  • Quick and easy to assemble.
  • Gamma stable.