Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, One-Piece AL, 3/8 ID, 5/8 OD
Catalog No. NC3852830
Encompass_Preferred
One-piece AL-series tubing retainer that secures flexible tubing to barbed fittings, providing 360° radial crimp and 360° compression for a leak-resistant connection.
  • One-piece design combines sleeve and collet for simplified assembly and consistent installation.
  • 360° radial crimp and compression for secure, leak-resistant seal on barbed fittings.
  • Designed for 0.375 in inner diameter (3/8 in) tubing and 0.625 in outer diameter (5/8 in) tubing.
  • Material options include polypropylene; PVDF option available per datasheet.
  • Suitable for biopharmaceutical fluid transfer, disposable assemblies, dialysis, and IV applications.
Catalog No. NC3852830 Supplier Accuflow Systems Supplier No. BL135375AL
One-piece AL-series tubing retainer that secures flexible tubing to barbed fittings, providing 360° radial crimp and 360° compression for a leak-resistant connection.
  • One-piece design combines sleeve and collet for simplified assembly and consistent installation.
  • 360° radial crimp and compression for secure, leak-resistant seal on barbed fittings.
  • Designed for 0.375 in inner diameter (3/8 in) tubing and 0.625 in outer diameter (5/8 in) tubing.
  • Material options include polypropylene; PVDF option available per datasheet.
  • Suitable for biopharmaceutical fluid transfer, disposable assemblies, dialysis, and IV applications.