Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, Polypropylene, 0.375 x 0.500 in., 250/Bag
Supplier:  Accuflow Systems BL135373AL
Encompass_Preferred
The BarbLock® retainer is an advanced product designed for retaining flexible tubing and hose onto barbed fittings, replacing inconsistent and leak-prone cable ties and traditional clamps. Its design incorporates a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge, effectively solving leak path problems in demanding fluid transfer applications.
  • Compatible with all flexible tubing materials
  • Provides a full 360° radial crimp behind the barb
  • Offers 360° compression directly on the barb's ridge
  • Eliminates the need for solvent bonding, ultraviolet bonding, and toxic adhesives
  • Manufactured from resins that meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements
  • Quick and easy to assemble
  • Ultra-secure tubing retainers
Catalog No. NC3935979