Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, Polypropylene, 0.500 in. ID, 250/Pk.
Catalog No. NC3936010
Encompass_Preferred
BarbLock® retainers securely attach flexible tubing to barbed fittings, providing a reliable alternative to traditional clamps. They feature a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge, preventing leaks in fluid transfer applications. This retainer is constructed from high-performance polypropylene.
  • Compatible with all flexible tubing materials.
  • Provides full 360° radial crimp behind barb and 360° compression directly on barb's ridge.
  • Eliminates need for solvent bonding and toxic adhesives.
  • Manufactured from resins meeting USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5 requirements.
  • Quick and easy to assemble.
  • Gamma stable.
Catalog No. NC3936010 Supplier Accuflow Systems Supplier No. BL135501
BarbLock® retainers securely attach flexible tubing to barbed fittings, providing a reliable alternative to traditional clamps. They feature a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge, preventing leaks in fluid transfer applications. This retainer is constructed from high-performance polypropylene.
  • Compatible with all flexible tubing materials.
  • Provides full 360° radial crimp behind barb and 360° compression directly on barb's ridge.
  • Eliminates need for solvent bonding and toxic adhesives.
  • Manufactured from resins meeting USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5 requirements.
  • Quick and easy to assemble.
  • Gamma stable.