Learn More
Accuflow Systems Saint-Gobain BarbLock Ultra-Secure Retainer, PVDF, 0.500 in. ID, 250/Pk.
Catalog No. NC3965648
Encompass
BarbLock® retainers secure flexible tubing and hose onto barbed fittings, offering a full 360° radial crimp behind the barb and 360° compression over the ridge for leak prevention. Made from high-performance PVDF resins, they meet USP <88> Class VI, USP <87>, and ISO 10993-5 requirements, suitable for sensitive liquids and gases.
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Offers 360° compression directly on the barb's ridge
  • Eliminates solvent bonding and toxic adhesives
  • Manufactured from USP Class VI resins
  • Quick and easy to assemble
Catalog No. NC3965648 Supplier Accuflow Systems Supplier No. BL135502PVDF
BarbLock® retainers secure flexible tubing and hose onto barbed fittings, offering a full 360° radial crimp behind the barb and 360° compression over the ridge for leak prevention. Made from high-performance PVDF resins, they meet USP <88> Class VI, USP <87>, and ISO 10993-5 requirements, suitable for sensitive liquids and gases.
  • Compatible with all flexible tubing materials
  • Provides 360° radial crimp behind the barb
  • Offers 360° compression directly on the barb's ridge
  • Eliminates solvent bonding and toxic adhesives
  • Manufactured from USP Class VI resins
  • Quick and easy to assemble