Learn More
Accuflow Systems Saint-Gobain BarbLock Ultra-Secure Tubing Retainer, PP, 0.125 ID, 0.250 OD
Supplier:  Accuflow Systems BL135125AL
Encompass_Preferred
The BarbLock® retainer is designed for securing flexible tubing onto barbed fittings. It features 360° radial crimping behind the barb and 360° compression directly over the barb's ridge, effectively addressing leak paths in demanding fluid transfer applications. This AL series one-piece retainer simplifies assembly by combining the sleeve and collet.
  • Compatible with all flexible tubing materials.
  • Full 360° radial crimp behind the barb.
  • Provides 360° compression directly on the barb's ridge.
  • Eliminates solvent bonding, ultraviolet bonding, and toxic adhesives.
  • Manufactured from resins meeting USP <88> Class VI, USP <87>, or ISO 10993-5.
  • Custom color and identification options available.
  • Quick and easy to assemble.
  • PP (polypropylene) and PVDF (polyvinylidene fluoride) retainers are gamma stable.
Catalog No. NC3935956