Learn More
Accuflow Systems Saint-Gobain Tygon A-60-G Tubing, 1/4 x 3/8 in., 50 ft.
Catalog No. NC3986081
Encompass_Preferred
Tygon® A-60-G Tubing is a high-performance chemical dispensing tubing designed for demanding industrial applications. It offers superior weathering, abrasion resistance, and an unequalled lifespan in peristaltic pump and vacuum systems.
  • Superior weathering
  • Abrasion resistant
  • Outstanding flexural fatigue resistance
  • Wide temperature range (-75°F to 275°F)
  • Low gas permeability versus rubber tubing
  • Ozone and UV light resistant
  • Designed to resist hydrocarbons
  • Engineered for outstanding performance and on-the-job reliability
  • Unequalled life in peristaltic pump applications
  • Ideal for use in vacuum systems
Catalog No. NC3986081 Supplier Accuflow Systems Supplier No. AFL00017
Tygon® A-60-G Tubing is a high-performance chemical dispensing tubing designed for demanding industrial applications. It offers superior weathering, abrasion resistance, and an unequalled lifespan in peristaltic pump and vacuum systems.
  • Superior weathering
  • Abrasion resistant
  • Outstanding flexural fatigue resistance
  • Wide temperature range (-75°F to 275°F)
  • Low gas permeability versus rubber tubing
  • Ozone and UV light resistant
  • Designed to resist hydrocarbons
  • Engineered for outstanding performance and on-the-job reliability
  • Unequalled life in peristaltic pump applications
  • Ideal for use in vacuum systems