Learn More
Accuflow Systems Saint-Gobain Peristaltic Pump Tubing, 5/8 In ID, 7/8 In OD
Catalog No. NC3856494
Encompass_Preferred
Saint-Gobain PharmaPure peristaltic pump tubing is a low-spallation thermoplastic tubing engineered for bioprocessing and peristaltic pump applications. This 25 ft coil in the 5/8 in ID x 7/8 in OD configuration provides consistent flow, extended pump life, and low particle generation for high-purity fluid transfer.
  • Low particle shedding for high-purity applications.
  • Thermoplastic construction designed for peristaltic pump compatibility.
  • 5/8 in ID, 7/8 in OD, 1/8 in wall, 25 ft coil.
  • Maximum working pressure 12 psi at 73 °F.
  • Vacuum rating 25 inHg at 73 °F.
  • Minimum bend radius 2-3/4 in.
Catalog No. NC3856494 Supplier Accuflow Systems Supplier No. AL242046
Saint-Gobain PharmaPure peristaltic pump tubing is a low-spallation thermoplastic tubing engineered for bioprocessing and peristaltic pump applications. This 25 ft coil in the 5/8 in ID x 7/8 in OD configuration provides consistent flow, extended pump life, and low particle generation for high-purity fluid transfer.
  • Low particle shedding for high-purity applications.
  • Thermoplastic construction designed for peristaltic pump compatibility.
  • 5/8 in ID, 7/8 in OD, 1/8 in wall, 25 ft coil.
  • Maximum working pressure 12 psi at 73 °F.
  • Vacuum rating 25 inHg at 73 °F.
  • Minimum bend radius 2-3/4 in.