Learn More
Pacer Global LLC Saint Gobain PharmaPure Tubing, 0.125'' ID, 0.25'' OD, 0.0625'' Wall, 25 Foot Coil, 1/EA
SDP
Catalog No. 5000332363
Encompass_Preferred
Saint-Gobain PharmaPure Pump Tubing, 0.125" ID, 0.250" OD, 0.0625" Wall, 25-Foot Coil, 1/EA
Catalog No. 50-003-32363 Supplier Pacer Global LLC Supplier No. MLSAL242007
Saint-Gobain PharmaPure Pump Tubing, 0.125" ID, 0.250" OD, 0.0625" Wall, 25-Foot Coil, 1/EA