Learn More
Pacer Global LLC Saint Gobain PharmaPure Tubing, 0.25'' ID, 0.375'' OD, 0.0625'' Wall, 25 Foot Coil, 1/EA
SDP
Catalog No. 5000332249
Encompass_Preferred
Saint-Gobain PharmaPure Pump Tubing, 0.250" ID, 0.375" OD, 0.0625" Wall, 25-Foot Coil, 1/EA
Catalog No. 50-003-32249 Supplier Pacer Global LLC Supplier No. MLSAL242017
Saint-Gobain PharmaPure Pump Tubing, 0.250" ID, 0.375" OD, 0.0625" Wall, 25-Foot Coil, 1/EA