Learn More
Accuflow Systems Saint-Gobain PharmaPure Tubing, 1/2 in x 3/4 in, 25 ft
Catalog No. NC3843997
Encompass_Preferred
Saint-Gobain PharmaPure peristaltic pump tubing provides ultra-low particulate spallation and extended pump life for pharmaceutical, biotechnology, and laboratory peristaltic pump applications. The proprietary multilayer thermoplastic elastomer construction offers low gas permeability and withstands common sterilization methods.
  • Low particulate spallation for extended pump life.
  • Proprietary multilayer thermoplastic elastomer construction.
  • 1/2 in inner diameter, 3/4 in outer diameter, 25 ft length.
  • 1/8 in wall thickness, opaque/white color.
  • Withstands EtO, autoclave, and gamma sterilization.
  • Working pressure 15 psi, maximum pressure 75 psi.
  • Meets USP Class VI and European Pharmacopoeia requirements where applicable.
Catalog No. NC3843997 Supplier Accuflow Systems Supplier No. AL242038
Saint-Gobain PharmaPure peristaltic pump tubing provides ultra-low particulate spallation and extended pump life for pharmaceutical, biotechnology, and laboratory peristaltic pump applications. The proprietary multilayer thermoplastic elastomer construction offers low gas permeability and withstands common sterilization methods.
  • Low particulate spallation for extended pump life.
  • Proprietary multilayer thermoplastic elastomer construction.
  • 1/2 in inner diameter, 3/4 in outer diameter, 25 ft length.
  • 1/8 in wall thickness, opaque/white color.
  • Withstands EtO, autoclave, and gamma sterilization.
  • Working pressure 15 psi, maximum pressure 75 psi.
  • Meets USP Class VI and European Pharmacopoeia requirements where applicable.