Learn More
Accuflow Systems Saint-Gobain PharmaPure Tubing, 1/8 in x 1/4 in, 25 ft
Catalog No. NC3856492
Encompass_Preferred
PharmaPure Low Spallation Peristaltic Pump Tubing is engineered for peristaltic pump applications requiring minimal particulate generation, extended pump life, and low gas permeability. The tubing is designed to maintain mechanical integrity through sterilization and gamma irradiation processes, making it suitable for sterile filling and bioprocess lines.
  • Ultra-low particulate spallation.
  • Outlasts silicone tubing in peristaltic pumps, extending pump life.
  • Excellent gas barrier with very low permeability.
  • Autoclavable and sterilizable.
  • Gamma stable with maintained performance up to 50 kGy.
  • 1/8 in (ID) x 1/4 in (OD), 25 ft coil.
  • Meets USP Biological Test for Plastics, Class VI; documented biocompatibility available on request.
  • Common applications: sterile filling, bioreactor process lines, media process systems, filtration, and shear-sensitive fluid transfer.
Catalog No. NC3856492 Supplier Accuflow Systems Supplier No. AL242007
PharmaPure Low Spallation Peristaltic Pump Tubing is engineered for peristaltic pump applications requiring minimal particulate generation, extended pump life, and low gas permeability. The tubing is designed to maintain mechanical integrity through sterilization and gamma irradiation processes, making it suitable for sterile filling and bioprocess lines.
  • Ultra-low particulate spallation.
  • Outlasts silicone tubing in peristaltic pumps, extending pump life.
  • Excellent gas barrier with very low permeability.
  • Autoclavable and sterilizable.
  • Gamma stable with maintained performance up to 50 kGy.
  • 1/8 in (ID) x 1/4 in (OD), 25 ft coil.
  • Meets USP Biological Test for Plastics, Class VI; documented biocompatibility available on request.
  • Common applications: sterile filling, bioreactor process lines, media process systems, filtration, and shear-sensitive fluid transfer.