Learn More
Pacer Global LLC Saint Gobain PharMed BPT Tubing, 0.02'' ID, 0.145'' OD, 0.0625'' Wall, 25 Foot Coil, 1/EA
SDP
Catalog No. 5000332369
Encompass_Preferred
Catalog No. 50-003-32369 Supplier Pacer Global LLC Supplier No. MLSAY242605