Learn More
Pacer Global LLC Saint-Gobain - PharMed BPT Tubing, 0.5'' ID, 0.75'' OD, 0.125'' Wall, 25 Foot Coil, 1/EA - AY242038
SDP
Catalog No. 5000332349
Encompass_Preferred
Saint-Gobain - PharMed BPT Tubing, 0.5" ID, 0.75" OD, 0.125" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32349 Supplier Pacer Global LLC Supplier No. MLSAY242038
Saint-Gobain - PharMed BPT Tubing, 0.5" ID, 0.75" OD, 0.125" Wall, 25 Foot Coil, 1/EA