Learn More
Accuflow Systems Saint-Gobain Sani-Tech SPT-60L Pump Tubing, 0.236 in. ID, 25 ft.
Catalog No. NC3953638
Encompass_Preferred
Sani-Tech SPT-60L is a premium platinum-cured silicone tubing designed for demanding pharmaceutical operations. It offers best-in-class accuracy and precision for consistent filling, minimizing costly over/under fills, and features low spallation and long pump life.
  • Long pump life (over 250 hours average)
  • Low spallation (50% less than closest competitor)
  • Accurate and consistent dispensing (4X improvement in accuracy, 2X in precision)
  • Full extractables report per BioPhorum Operations Group (BPOG) protocol
  • Technical dossier available
  • Available with ValPlus™ certification
  • Manufactured from platinum-cured silicone, tested to USP <88> Class VI, USP <87>, ISO 10993-5, and EP 3.1.9
  • Biocompatible with ADCF/BSE/TSE statements
  • Sterilization methods: autoclavable, gamma irradiation, and gas (ethylene oxide)
Catalog No. NC3953638 Supplier Accuflow Systems Supplier No. SPT60L236394
Sani-Tech SPT-60L is a premium platinum-cured silicone tubing designed for demanding pharmaceutical operations. It offers best-in-class accuracy and precision for consistent filling, minimizing costly over/under fills, and features low spallation and long pump life.
  • Long pump life (over 250 hours average)
  • Low spallation (50% less than closest competitor)
  • Accurate and consistent dispensing (4X improvement in accuracy, 2X in precision)
  • Full extractables report per BioPhorum Operations Group (BPOG) protocol
  • Technical dossier available
  • Available with ValPlus™ certification
  • Manufactured from platinum-cured silicone, tested to USP <88> Class VI, USP <87>, ISO 10993-5, and EP 3.1.9
  • Biocompatible with ADCF/BSE/TSE statements
  • Sterilization methods: autoclavable, gamma irradiation, and gas (ethylene oxide)