Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.03'' ID, 0.155'' OD, 0.062'' Wall, 25 Foot Coil, 1/EA - STHT-C-030-2
SDP
Catalog No. 5000332279
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.03" ID, 0.155" OD, 0.062" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32279 Supplier Pacer Global LLC Supplier No. MLSSTHTC0302
Saint-Gobain - SaniTech-C Tubing, 0.03" ID, 0.155" OD, 0.062" Wall, 25 Foot Coil, 1/EA