Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.09375'' ID, 0.15625'' OD, 0.03125'' Wall, 100 Foot Coil, 1/EA - STHT-C-093-1H
SDP
Catalog No. 5000332415
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.09375" ID, 0.15625" OD, 0.03125" Wall, 100 Foot Coil, 1/EA
Catalog No. 50-003-32415 Supplier Pacer Global LLC Supplier No. MLSSTHTC0931H
Saint-Gobain - SaniTech-C Tubing, 0.09375" ID, 0.15625" OD, 0.03125" Wall, 100 Foot Coil, 1/EA