Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.09375'' ID, 0.21875'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - STHT-C-093-2F
SDP
Catalog No. 5000332416
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.09375" ID, 0.21875" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32416 Supplier Pacer Global LLC Supplier No. MLSSTHTC0932F
Saint-Gobain - SaniTech-C Tubing, 0.09375" ID, 0.21875" OD, 0.0625" Wall, 50 Foot Coil, 1/EA