Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.125'' ID, 0.1875'' OD, 0.03125'' Wall, 25 Foot Coil, 1/EA - STHT-C-125-1
SDP
Catalog No. 5000332477
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.125" ID, 0.1875" OD, 0.03125" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32477 Supplier Pacer Global LLC Supplier No. MLSSTHTC1251
Saint-Gobain - SaniTech-C Tubing, 0.125" ID, 0.1875" OD, 0.03125" Wall, 25 Foot Coil, 1/EA